siRNA Details
| Gene Symbol: WFDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145652.3 | NP_663627.1 | 2 | 205 | ||
| XM_011528601.1 | XP_011526903.1 | 3 | 706 | |||
| XM_011528602.1 | XP_011526904.1 | 4 | 514 | |||
| GenBank mRNA | AY038181.1 | - | 2 | 165 | ||
| AY358822.1 | - | 2 | 205 | |||
| BC039173.1 | - | 2 | 171 | 4755260 | ||

WFDC5
Genomic Map