siRNA Details
Gene Symbol: LINC00654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015406.2 | 2 | 1576 | |||
GenBank mRNA | AL137476.1 | - | 1 | 716 | ||
BC067900.1 | - | 2 | 1593 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015406.2 | 2 | 1576 | |||
GenBank mRNA | AL137476.1 | - | 1 | 716 | ||
BC067900.1 | - | 2 | 1593 |