siRNA Details
| Gene Symbol: LINC00654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015406.2 | 2 | 1655 | |||
| GenBank mRNA | AL137476.1 | - | 1 | 795 | ||
| BC067900.1 | - | 2 | 1672 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00654| Gene Symbol: LINC00654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015406.2 | 2 | 1655 | |||
| GenBank mRNA | AL137476.1 | - | 1 | 795 | ||
| BC067900.1 | - | 2 | 1672 | |||