siRNA Details
| Gene Symbol: ZNF295-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027273.2 | 2 | 291 | |||
| NR_119384.1 | 2 | 520 | ||||
| NR_119385.1 | 2 | 337 | ||||
| GenBank mRNA | AK055167.1 | - | 1 | 2351 | ||
| BC029588.1 | - | 2 | 291 | |||
| BI464310.1 | - | 2 | 325 | |||
| DB455362.1 | - | 2 | 337 | |||
| HY004634.1 | - | 2 | 321 | |||

ZNF295-AS1
Genomic Map