siRNA Details
| Gene Symbol: CHADL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138481.1 | NP_612490.1 | 5 | 2295 | ||
| XM_005261370.3 | XP_005261427.1 | 5 | 2341 | |||
| XM_011529932.2 | XP_011528234.1 | 4 | 2959 | |||
| XM_011529933.2 | XP_011528235.1 | 4 | 2957 | |||
| XM_011529934.2 | XP_011528236.1 | 4 | 2959 | |||
| GenBank mRNA | BC012882.1 | - | 2 | 342 | ||
| BC019839.1 | - | 3 | 1482 | |||
| BC040188.1 | - | 5 | 2035 | |||
| BC048421.1 | - | 3 | 1318 | |||
| BC068590.1 | - | 5 | 2295 | |||

CHADL
Genomic Map