siRNA Details
| Gene Symbol: FAM109B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002034.2 | NP_001002034.2 | 3 | 583 | ||
| XM_005261373.3 | XP_005261430.1 | 3 | 721 | |||
| GenBank mRNA | BC029776.1 | - | 1 | 237 | ||
| BC104175.1 | - | 2 | 479 | 40030677 | ||
| BC104176.1 | - | 2 | 477 | 40030679 | ||
| BX648402.1 | - | 3 | 583 | |||
| CR456454.1 | - | 2 | 456 | |||

FAM109B
Genomic Map