siRNA Details
| Gene Symbol: LINC01104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103730.1 | 1, 2 | 564 | |||
| GenBank mRNA | AK056640.1 | - | 1, 2 | 564 | ||
| BC105017.1 | - | 1, 2 | 168 | |||
| BC105019.1 | - | 1, 2 | 168 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01104| Gene Symbol: LINC01104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103730.1 | 1, 2 | 564 | |||
| GenBank mRNA | AK056640.1 | - | 1, 2 | 564 | ||
| BC105017.1 | - | 1, 2 | 168 | |||
| BC105019.1 | - | 1, 2 | 168 | |||