siRNA Details
Gene Symbol: LOC151121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122040.1 | 5 | 1855 | |||
NR_122041.1 | 4 | 1767 | ||||
NR_122042.1 | 4 | 1778 | ||||
GenBank mRNA | AK056598.1 | - | 5 | 1812 | ||
BC093715.1 | - | 1 | 213 | 7939560 | ||
BC111988.1 | - | 1 | 213 | 8327456 |
