siRNA Details
| Gene Symbol: LOC151121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122040.1 | 5 | 1921 | |||
| NR_122041.1 | 4 | 1833 | ||||
| NR_122042.1 | 4 | 1844 | ||||
| GenBank mRNA | AK056598.1 | - | 5 | 1878 | ||
| BC093715.1 | - | 1 | 279 | 7939560 | ||
| BC111988.1 | - | 1 | 279 | 8327456 | ||

LOC151121
Genomic Map