siRNA Details
| Gene Symbol: MIR3681HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110197.1 | 3 | 289 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR3681HG| Gene Symbol: MIR3681HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110197.1 | 3 | 289 | |||