siRNA Details
| Gene Symbol: LINC00608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015390.1 | 1 | 329 | |||
| NR_024385.1 | 1 | 329 | ||||
| GenBank mRNA | AK098539.1 | - | 1 | 329 | ||
| BC033552.1 | - | 1 | 311 | |||
| BC049373.1 | - | 1 | 329 | |||
| BC121084.1 | - | 1 | 311 | |||
| BC121085.1 | - | 1 | 311 | |||

LINC00608
Genomic Map