siRNA Details
| Gene Symbol: LOC101928327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130782.1 | 2 | 324 | |||
| GenBank mRNA | BC021184.1 | - | 2 | 329 | ||
| BC090895.1 | - | 2 | 297 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928327| Gene Symbol: LOC101928327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130782.1 | 2 | 324 | |||
| GenBank mRNA | BC021184.1 | - | 2 | 329 | ||
| BC090895.1 | - | 2 | 297 | |||