siRNA Details
| Gene Symbol: LINC00635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015414.1 | 8 | 757 | |||
| GenBank mRNA | BC045586.1 | - | 8 | 757 | ||
| BC104745.1 | - | 4 | 351 | |||
| BC104746.1 | - | 3 | 296 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00635| Gene Symbol: LINC00635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015414.1 | 8 | 757 | |||
| GenBank mRNA | BC045586.1 | - | 8 | 757 | ||
| BC104745.1 | - | 4 | 351 | |||
| BC104746.1 | - | 3 | 296 | |||