siRNA Details
| Gene Symbol: ZCWPW2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040432.3 | NP_001035522.1 | 10 | 1167 | ||
| NM_001324169.1 | NP_001311098.1 | 9 | 1047 | |||
| NM_001324170.1 | NP_001311099.1 | 7 | 861 | |||
| NR_136708.1 | 8 | 929 | ||||
| XM_017005755.1 | XP_016861244.1 | 9 | 4842 | |||
| XM_017005756.1 | XP_016861245.1 | 7 | 4659 | |||
| GenBank mRNA | BC065764.1 | - | 8 | 635 | ||
| BC094696.1 | - | 10 | 1147 | 30529418 | ||

ZCWPW2
Genomic Map