siRNA Details
| Gene Symbol: FAM53A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013622.3 | NP_001013644.1 | 4 | 450 | ||
| NM_001174070.2 | NP_001167541.1 | 4 | 497 | |||
| NM_001297435.1 | NP_001284364.1 | 4 | 850 | |||
| XM_005247948.3 | XP_005248005.1 | 4 | 1147 | |||
| XM_011513402.2 | XP_011511704.1 | 4 | 1153 | |||
| XM_011513403.1 | XP_011511705.1 | 4 | 1147 | |||
| XM_011513404.2 | XP_011511706.1 | 4 | 1147 | |||
| XM_011513405.1 | XP_011511707.1 | 4 | 1147 | |||
| XM_017007802.1 | XP_016863291.1 | 4 | 1147 | |||
| GenBank mRNA | AK125566.1 | - | 4 | 445 | ||
| BC070112.1 | - | 4 | 450 | 5268061 | ||

FAM53A
Genomic Map