siRNA Details
| Gene Symbol: LOC100505841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195535.1 | NP_001182464.1 | 3 | 335 | ||
| XM_011543086.2 | XP_011541388.1 | 6 | 654 | |||
| GenBank mRNA | AI188208.1 | - | 1 | 106 | ||
| BU567915.1 | - | 3 | 338 | |||

LOC100505841
Genomic Map