siRNA Details
Gene Symbol: PPP1R2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038443.1 | 1 | 894 | |||
GenBank mRNA | BC066922.1 | - | 1 | 894 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PPP1R2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038443.1 | 1 | 894 | |||
GenBank mRNA | BC066922.1 | - | 1 | 894 |