siRNA Details
| Gene Symbol: LINC01010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038216.1 | 1 | 122 | |||
| NR_038217.1 | 1 | 122 | ||||
| GenBank mRNA | AK055166.1 | - | 1 | 125 | ||
| DA419376.1 | - | 1 | 125 | |||
| DA420003.1 | - | 1 | 124 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01010| Gene Symbol: LINC01010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038216.1 | 1 | 122 | |||
| NR_038217.1 | 1 | 122 | ||||
| GenBank mRNA | AK055166.1 | - | 1 | 125 | ||
| DA419376.1 | - | 1 | 125 | |||
| DA420003.1 | - | 1 | 124 | |||