siRNA Details
| Gene Symbol: KHDC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017361.2 | NP_001017361.1 | 1 | 80 | ||
| GenBank mRNA | AB211062.1 | - | 1 | 80 | ||
| BC132844.1 | - | 1 | 72 | 40146866 | ||
| BC137160.1 | - | 1 | 72 | 9021157 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KHDC3L| Gene Symbol: KHDC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017361.2 | NP_001017361.1 | 1 | 80 | ||
| GenBank mRNA | AB211062.1 | - | 1 | 80 | ||
| BC132844.1 | - | 1 | 72 | 40146866 | ||
| BC137160.1 | - | 1 | 72 | 9021157 | ||