siRNA Details
| Gene Symbol: C7orf77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024603.1 | NP_001019774.1 | 4 | 430 | ||
| GenBank mRNA | BC130600.1 | - | 4 | 309 | ||
| BC130604.1 | - | 4 | 309 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7orf77| Gene Symbol: C7orf77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024603.1 | NP_001019774.1 | 4 | 430 | ||
| GenBank mRNA | BC130600.1 | - | 4 | 309 | ||
| BC130604.1 | - | 4 | 309 | |||