siRNA Details
| Gene Symbol: SBDSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001588.2 | 4 | 799 | |||
| NR_024109.1 | 3 | 677 | ||||
| NR_024110.1 | 4 | 799 | ||||
| NR_024111.1 | 3 | 677 | ||||
| GenBank mRNA | BQ441749.1 | - | 3 | 536 | ||
| CB216674.1 | - | 2 | 228 | |||
| CN267170.1 | - | 3 | 562 | |||
| CX867149.1 | - | 4 | 691 | |||

SBDSP1
Genomic Map