siRNA Details
| Gene Symbol: AGR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176813.4 | NP_789783.1 | 6, 7 | 435 | ||
| XM_005249627.2 | XP_005249684.1 | 6, 7 | 414 | |||
| XM_011515152.2 | XP_011513454.1 | 6, 7 | 427 | |||
| GenBank mRNA | AY069977.1 | - | 5, 6 | 400 | ||
| AY359065.1 | - | 6, 7 | 419 | |||
| BC058284.1 | - | 6, 7 | 440 | 4694757 | ||
| BG540617.1 | - | 439 | ||||
| BM984550.1 | - | 2, 3 | 323 | |||
| BU682778.1 | - | 2, 3 | 329 | |||
| CB242846.1 | - | 2, 3 | 323 | |||

AGR3
Genomic Map