siRNA Details
| Gene Symbol: FAM87A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103537.1 | 4 | 2985 | |||
| GenBank mRNA | BC037297.2 | - | 4 | 2984 | ||
| BC118644.1 | - | 1 | 801 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM87A| Gene Symbol: FAM87A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103537.1 | 4 | 2985 | |||
| GenBank mRNA | BC037297.2 | - | 4 | 2984 | ||
| BC118644.1 | - | 1 | 801 | |||