siRNA Details
Gene Symbol: FAM87A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103537.1 | 4 | 3132 | |||
GenBank mRNA | BC037297.2 | - | 4 | 3131 | ||
BC118644.1 | - | 1 | 948 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM87A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103537.1 | 4 | 3132 | |||
GenBank mRNA | BC037297.2 | - | 4 | 3131 | ||
BC118644.1 | - | 1 | 948 |