siRNA Details
Gene Symbol: LINC00475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027341.1 | 4 | 506 | |||
GenBank mRNA | AK023662.1 | - | 4 | 506 | ||
BC128384.1 | - | 5 | 574 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027341.1 | 4 | 506 | |||
GenBank mRNA | AK023662.1 | - | 4 | 506 | ||
BC128384.1 | - | 5 | 574 |