siRNA Details
| Gene Symbol: ATP8B5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003582.1 | 11 | 1354 | |||
| NR_110320.1 | 11 | 1350 | ||||
| GenBank mRNA | BC035229.1 | - | 10 | 2760 | ||
| BC130448.1 | - | 6 | 1114 | |||
| BC130450.1 | - | 6 | 961 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP8B5P| Gene Symbol: ATP8B5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003582.1 | 11 | 1354 | |||
| NR_110320.1 | 11 | 1350 | ||||
| GenBank mRNA | BC035229.1 | - | 10 | 2760 | ||
| BC130448.1 | - | 6 | 1114 | |||
| BC130450.1 | - | 6 | 961 | |||