siRNA Details
| Gene Symbol: CSAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BF570999.1 | - | 4 | 414 | ||
| BM810082.1 | - | 4 | 396 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSAG1| Gene Symbol: CSAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BF570999.1 | - | 4 | 414 | ||
| BM810082.1 | - | 4 | 396 | |||