siRNA Details
| Gene Symbol: SMIM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162936.1 | NP_001156408.1 | 3 | 327 | ||
| GenBank mRNA | BX092805.1 | - | 2 | 215 | ||
| DA855214.1 | - | 3 | 327 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM9| Gene Symbol: SMIM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162936.1 | NP_001156408.1 | 3 | 327 | ||
| GenBank mRNA | BX092805.1 | - | 2 | 215 | ||
| DA855214.1 | - | 3 | 327 | |||