siRNA Details
| Gene Symbol: REM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173527.2 | NP_775798.2 | 2 | 477 | ||
| XM_005267383.3 | XP_005267440.1 | 2 | 535 | |||
| XM_011536518.2 | XP_011534820.1 | 1 | 1684 | |||
| XM_017021058.1 | XP_016876547.1 | 1 | 1684 | |||
| XM_017021059.1 | XP_016876548.1 | 1 | 1684 | |||
| GenBank mRNA | AK096283.1 | - | 2 | 477 | ||
| AK299238.1 | - | 2 | 478 | |||
| BC035663.1 | - | 2 | 468 | 5723443 | ||
| KU178879.1 | - | 2 | 382 | |||
| KU178880.1 | - | 2 | 382 | |||
| KU178881.1 | - | 2 | 382 | |||

REM2
Genomic Map