siRNA Details
| Gene Symbol: SLC16A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153357.1 | NP_699188.1 | 2 | 694 | ||
| XM_005256488.3 | XP_005256545.2 | 3 | 927 | |||
| XM_017024281.1 | XP_016879770.1 | 3 | 927 | |||
| XM_017024282.1 | XP_016879771.1 | 3 | 927 | |||
| GenBank mRNA | AK074674.1 | - | 2 | 694 | ||
| BC093860.1 | - | 2 | 377 | 7939705 | ||
| BC093886.1 | - | 2 | 377 | 7939731 | ||
| BC143369.1 | - | 1 | 615 | |||

SLC16A11
Genomic Map