siRNA Details
| Gene Symbol: ZNF497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207009.1 | NP_001193938.1 | 2 | 1545 | ||
| NM_198458.2 | NP_940860.2 | 3 | 1642 | |||
| GenBank mRNA | AK126727.1 | - | 3 | 1548 | ||
| AK131209.1 | - | 2 | 1545 | |||
| BC118983.1 | - | 2 | 1410 | 40025731 | ||
| BC125273.1 | - | 2 | 1415 | 40025732 | ||

ZNF497
Genomic Map