siRNA Details
| Gene Symbol: ZNF497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207009.1 | NP_001193938.1 | 2 | 1623 | ||
| NM_198458.2 | NP_940860.2 | 3 | 1720 | |||
| GenBank mRNA | AK126727.1 | - | 3 | 1626 | ||
| AK131209.1 | - | 2 | 1623 | |||
| BC118983.1 | - | 2 | 1488 | 40025731 | ||
| BC125273.1 | - | 2 | 1493 | 40025732 | ||
| Gene Symbol: A1BG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC035719.1 | - | 1 | 27 | 5588885 | |

ZNF497
Genomic Map