siRNA Details
| Gene Symbol: ZNF709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152601.3 | NP_689814.1 | 4 | 369 | ||
| GenBank mRNA | AK290911.1 | - | 4 | 308 | ||
| BC136746.1 | - | 4 | 285 | 9020736 | ||
| BC136751.1 | - | 4 | 285 | 9020741 | ||
| BX954970.1 | - | 4 | 370 | |||
| DA493682.1 | - | 4 | 350 | |||

ZNF709
Genomic Map