siRNA Details
| Gene Symbol: PRR22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134316.1 | NP_001127788.1 | 3 | 1164 | ||
| GenBank mRNA | BC008985.1 | - | 1 | 1412 | 4213313 | |
| BC023278.1 | - | 1 | 1412 | 4213314 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR22| Gene Symbol: PRR22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134316.1 | NP_001127788.1 | 3 | 1164 | ||
| GenBank mRNA | BC008985.1 | - | 1 | 1412 | 4213313 | |
| BC023278.1 | - | 1 | 1412 | 4213314 | ||