siRNA Details
| Gene Symbol: ZNF737 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159293.1 | NP_001152765.1 | 2 | 197 | ||
| XM_005259697.3 | XP_005259754.1 | 2 | 186 | |||
| XM_006722597.3 | XP_006722660.1 | 2 | 186 | |||
| XM_011527598.2 | XP_011525900.1 | 2 | 186 | |||
| XM_011527599.2 | XP_011525901.1 | 2 | 186 | |||
| XM_017026116.1 | XP_016881605.1 | 3 | 284 | |||
| GenBank mRNA | DA043454.1 | - | 2 | 197 | ||

ZNF737
Genomic Map