siRNA Details
| Gene Symbol: CST13P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001279.2 | 4 | 967 | |||
| GenBank mRNA | BC024006.1 | - | 4 | 972 | ||
| BC028368.1 | - | 4 | 972 | |||
| BC033934.1 | - | 4 | 962 | |||
| BC041171.1 | - | 4 | 946 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CST13P| Gene Symbol: CST13P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001279.2 | 4 | 967 | |||
| GenBank mRNA | BC024006.1 | - | 4 | 972 | ||
| BC028368.1 | - | 4 | 972 | |||
| BC033934.1 | - | 4 | 962 | |||
| BC041171.1 | - | 4 | 946 | |||