siRNA Details
| Gene Symbol: FREM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168235.1 | NP_001161707.1 | 6 | 6010 | ||
| GenBank mRNA | AI184955.1 | - | 2 | 155 | ||
| DR731220.1 | - | 2 | 111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FREM3| Gene Symbol: FREM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168235.1 | NP_001161707.1 | 6 | 6010 | ||
| GenBank mRNA | AI184955.1 | - | 2 | 155 | ||
| DR731220.1 | - | 2 | 111 | |||