siRNA Details
| Gene Symbol: ZNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073389.1 | 3 | 1405 | |||
| GenBank mRNA | AL834121.1 | - | 1 | 922 | ||
| BC017945.1 | - | 1 | 265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF138| Gene Symbol: ZNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073389.1 | 3 | 1405 | |||
| GenBank mRNA | AL834121.1 | - | 1 | 922 | ||
| BC017945.1 | - | 1 | 265 | |||