siRNA Details
| Gene Symbol: TMEM252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153237.1 | NP_694969.1 | 2 | 492 | ||
| GenBank mRNA | BC029780.1 | - | 2 | 492 | 5185747 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM252| Gene Symbol: TMEM252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153237.1 | NP_694969.1 | 2 | 492 | ||
| GenBank mRNA | BC029780.1 | - | 2 | 492 | 5185747 | |