siRNA Details
Gene Symbol: TMEM252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153237.1 | NP_694969.1 | 1 | 80 | ||
GenBank mRNA | BC029780.1 | - | 1 | 80 | 5185747 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153237.1 | NP_694969.1 | 1 | 80 | ||
GenBank mRNA | BC029780.1 | - | 1 | 80 | 5185747 |