siRNA Details
Gene Symbol: LINC00858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038220.1 | 6 | 1409 | |||
GenBank mRNA | AK056561.1 | - | 6 | 1409 | ||
BC104895.1 | - | 1 | 63 | |||
BC104921.1 | - | 1 | 62 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038220.1 | 6 | 1409 | |||
GenBank mRNA | AK056561.1 | - | 6 | 1409 | ||
BC104895.1 | - | 1 | 63 | |||
BC104921.1 | - | 1 | 62 |