siRNA Details
| Gene Symbol: LINC00858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038220.1 | 6 | 1488 | |||
| GenBank mRNA | AK056561.1 | - | 6 | 1488 | ||
| BC104895.1 | - | 1 | 142 | |||
| BC104921.1 | - | 1 | 141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00858| Gene Symbol: LINC00858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038220.1 | 6 | 1488 | |||
| GenBank mRNA | AK056561.1 | - | 6 | 1488 | ||
| BC104895.1 | - | 1 | 142 | |||
| BC104921.1 | - | 1 | 141 | |||