siRNA Details
| Gene Symbol: ANKRD24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133475.1 | NP_597732.1 | 8 | 786 | ||
| XM_011527756.2 | XP_011526058.1 | 9 | 696 | |||
| XM_011527757.2 | XP_011526059.1 | 8 | 944 | |||
| XM_011527758.2 | XP_011526060.1 | 8 | 609 | |||
| XM_011527761.1 | XP_011526063.1 | 10 | 766 | |||
| XM_011527764.2 | XP_011526066.1 | 7 | 857 | |||
| XM_011527765.2 | XP_011526067.1 | 7 | 510 | |||
| XM_017026438.1 | XP_016881927.1 | 10 | 757 | |||
| XM_017026439.1 | XP_016881928.1 | 9 | 679 | |||
| XM_017026440.1 | XP_016881929.1 | 8 | 652 | |||
| GenBank mRNA | AB075861.1 | - | 6 | 483 | ||
| BC028236.1 | - | 6 | 428 | |||
| BI914435.1 | - | 6 | 480 | |||

ANKRD24
Genomic Map