siRNA Details
| Gene Symbol: FAM9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_205849.3 | NP_995321.1 | 4 | 526 | ||
| XM_011545464.2 | XP_011543766.1 | 11 | 1099 | |||
| XM_011545465.2 | XP_011543767.1 | 3 | 458 | |||
| GenBank mRNA | AF494344.1 | - | 4 | 525 | ||
| AK097501.1 | - | 3 | 485 | |||
| AK128470.1 | - | 5 | 1118 | |||
| BC120955.1 | - | 3 | 225 | 40115245 | ||
| BC120956.1 | - | 3 | 225 | 40115246 | ||

FAM9B
Genomic Map