siRNA Details
| Gene Symbol: AAED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153698.1 | NP_714542.1 | 2, 3 | 291 | ||
| XM_005251783.3 | XP_005251840.1 | 2, 3 | 286 | |||
| XM_005251784.4 | XP_005251841.1 | 2, 3 | 143 | |||
| GenBank mRNA | BC136501.1 | - | 2, 3 | 129 | 9020490 | |
| BC136503.1 | - | 2, 3 | 291 | 9020492 | ||
| BF445089.1 | - | 2, 3 | 166 | |||
| BM843088.1 | - | 2, 3 | 291 | |||

AAED1
Genomic Map