siRNA Details
| Gene Symbol: C12orf77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101339.2 | NP_001094809.2 | 3, 4 | 613 | ||
| XM_017018966.1 | XP_016874455.1 | 4, 5 | 624 | |||
| GenBank mRNA | BC035880.1 | - | 3, 4 | 624 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12orf77| Gene Symbol: C12orf77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101339.2 | NP_001094809.2 | 3, 4 | 613 | ||
| XM_017018966.1 | XP_016874455.1 | 4, 5 | 624 | |||
| GenBank mRNA | BC035880.1 | - | 3, 4 | 624 | ||