siRNA Details
| Gene Symbol: DNM1P46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003260.1 | 3 | 446 | |||
| GenBank mRNA | AK127834.1 | - | 3 | 446 | ||
| AK302717.1 | - | 2 | 374 | |||
| AL833912.1 | - | 2 | 298 | |||
| BC101078.3 | - | 2 | 294 | |||
| BC101080.2 | - | 3 | 334 | |||

DNM1P46
Genomic Map