siRNA Details
Gene Symbol: MGC15885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026897.1 | 2 | 820 | |||
GenBank mRNA | BC014404.1 | - | 2 | 942 | ||
BC131619.1 | - | 2 | 719 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MGC15885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026897.1 | 2 | 820 | |||
GenBank mRNA | BC014404.1 | - | 2 | 942 | ||
BC131619.1 | - | 2 | 719 |