siRNA Details
| Gene Symbol: TMEM201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010866.3 | NP_001010866.1 | 6 | 1222 | ||
| XM_011540910.2 | XP_011539212.1 | 6 | 1251 | |||
| GenBank mRNA | BC121187.1 | - | 6 | 1177 | 3870153 | |
| BC137291.1 | - | 6 | 1222 | 9021288 | ||

TMEM201
Genomic Map