siRNA Details
| Gene Symbol: C1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012970.2 | NP_001012988.1 | 4 | 414 | ||
| NM_001276348.1 | NP_001263277.1 | 4 | 318 | |||
| XM_011544124.2 | XP_011542426.1 | 5 | 1256 | |||
| XM_011544125.2 | XP_011542427.1 | 4 | 1006 | |||
| XM_011544128.2 | XP_011542430.1 | 4 | 1517 | |||
| XM_011544129.2 | XP_011542431.1 | 5 | 1167 | |||
| XM_017000571.1 | XP_016856060.1 | 5 | 1286 | |||
| XM_017000572.1 | XP_016856061.1 | 5 | 1794 | |||
| GenBank mRNA | BC054479.1 | - | 4 | 322 | ||
| HY055397.1 | - | 4 | 370 | |||

C1orf100
Genomic Map